Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Bis(Trimethylsilyl)Amide Solution

Catalog No. 80631500MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-631-500MLF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 80-631-500MLF Supplier Sigma Aldrich Supplier No. 80631500MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium bis(trimethylsilyl)amide, also known as NaHMDS, is a metal dialkylamide reagent that combines both high basicity and nucleophilicity. It is a general reagent used for the enolization of carbonyl compounds.It can be used to prepare phosphonium ylides in Wittig reaction.NaHMDS solution can also be used as a source of nitrogen in aminomethylation and in the synthesis of primary amines from the corresponding alkyl halides.prices for bulk quantities on request

Specifications

CAS 7772-79-4
Melting Point 186°C to 189°C
Molecular Formula C16H23NO10
Linear Formula C16H23NO10
MDL Number MFCD00006595
Quantity 5 g
Synonym beta-D-Glucosamine pentaacetate; 2-Amino-2-deoxy-beta-D-glucopyranosyl pentaacetate
Molecular Weight (g/mol) 389.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.