accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7773-01-5

IUPAC Name: manganese(2+) dichloride
Synonyms: manganese(II) chloride
Molecular Weight (g/mol): 125.84
Molecular Formula: Cl2Mn
InChi Key: GLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Mn++]
Molecular Weight (g/mol) 
  • (20)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (5)
  • (3)
  • (4)
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (1)
  • (4)
  • (6)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (20)
  • (1)
  • (20)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (6)
  • (2)
  • (1)
  • (4)
  • (6)
  • (2)