Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) chloride, ultra dry, 99.99% (metals basis)

Catalog No. AA1446422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1446422 100 g
AA1446406 5 g
AA1446414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1446422 Supplier Thermo Scientific Chemicals Supplier No. 01446422
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol

Manganese(II) chloride is used in formation of an active Mn(0) species useful for radical cyclization reactions. It is used as catalyst, raw material for paint drier and antiknock agent for gasoline. It iis also used in the production of pigments, dyes, car batteries, chemical reagents and trace elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-01-5
Molecular Formula Cl2Mn
Molecular Weight (g/mol) 125.84
MDL Number MFCD00011114
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
PubChem CID 10313134
ChEBI CHEBI:63041
SMILES [Cl-].[Cl-].[Mn++]

Specifications

Boiling Point 1190°C
Melting Point 650°C
Color Pink
Physical Form Beads
Quantity 100 g
Linear Formula MnCl2
UN Number UN3288
Merck Index 14,5728
Solubility Information Soluble in water(723g/L at 25 C)
IUPAC Name manganese(2+) dichloride
Formula Weight 125.84
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 2.977 g/mL
Assay (metals basis)
Chemical Name or Material Manganese(II) chloride, Ultra dry
Show More Show Less
Hazard Category H302-H318-H373
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P262-P264-P270-P280i-P301+P312-P305+P351+P338-P310-P314-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-869-6
RTECSNumber OO9625000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.