CAS RN 7773-83-3
IUPAC Name:
docosane-1-thiol
Synonyms:
docosane-1-thiol
Molecular Weight (g/mol):
342.67
Molecular Formula:
C22H46S
InChi Key:
NNZMLOHQRXHPOZ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCCCS
1
–
1
of
1
results
1
1-Docosanethiol 98.0+%, TCI America™
CAS: 7773-83-3 Molecular Formula: C22H46S Molecular Weight (g/mol): 342.67 MDL Number: MFCD02262163 InChI Key: NNZMLOHQRXHPOZ-UHFFFAOYSA-N Synonym: Docosyl Mercaptan PubChem CID: 263213 IUPAC Name: docosane-1-thiol SMILES: CCCCCCCCCCCCCCCCCCCCCCS