Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Docosanethiol 98.0+%, TCI America™
SDP

Catalog No. D39055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D39055G Supplier TCI America Supplier No. D39055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Organic compound suitable for laboratory research applications.

Chemical Identifiers

CAS 7773-83-3
Molecular Formula C22H46S
Molecular Weight (g/mol) 342.67
MDL Number MFCD02262163
InChI Key NNZMLOHQRXHPOZ-UHFFFAOYSA-N
Synonym Docosyl Mercaptan
PubChem CID 263213
IUPAC Name docosane-1-thiol
SMILES CCCCCCCCCCCCCCCCCCCCCCS

Specifications

Melting Point 48°C
Color White
Quantity 5 g
Formula Weight 342.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Docosanethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.