accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7775-14-6

IUPAC Name: disodium sulfinatosulfinate
Synonyms: sodium dithionite sodium hydrosulfite
Molecular Weight (g/mol): 174.1
Molecular Formula: Na2O4S2
InChi Key: JVBXVOWTABLYPX-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)S([O-])=O
Grade 
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (7)
  • (2)
  • (2)
  • (3)
Physical Form 
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (7)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (3)