Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dithionite (Sodium Hydrosulfite), Reagents

Catalog No. 01337023
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-023 2.5 kg
01-337-022 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-023 Supplier Reagents Holdings Llc Supplier No. C23163002.5A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Dithionite (Sodium Hydrosulfite), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7775-14-6
Molecular Formula Na2O4S2
Molecular Weight (g/mol) 174.10
InChI Key JVBXVOWTABLYPX-UHFFFAOYSA-L
Synonym Sodium Hydrosulfite
IUPAC Name disodium sulfinatosulfinate
SMILES [Na+].[Na+].[O-]S(=O)S([O-])=O

Specifications

Boiling Point >80°C
Melting Point 52°C
Color White to Off-White
pH 8 to 10.5
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.19
Chemical Name or Material Sodium Dithionite (Sodium Hydrosulfite)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.