accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77758-79-3

IUPAC Name: (1,1,2,2,3,3,3-heptafluoropropyl)(phenyl)iodanium trifluoromethanesulfonate
Synonyms: (1,1,2,2,3,3,3-heptafluoropropyl)(phenyl)iodanium triflate
Molecular Weight (g/mol): 522.1
Molecular Formula: C10H5F10IO3S
InChi Key: YBVOPUOUKFHORP-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.FC(F)(F)C(F)(F)C(F)(F)[I+]C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results