Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Perfluoropropyl)phenyliodonium Trifluoromethanesulfonate 97.0+%, TCI America™
SDP

Catalog No. P10821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10821G Supplier TCI America Supplier No. P10821G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77758-79-3
Molecular Formula C10H5F10IO3S
Molecular Weight (g/mol) 522.096
MDL Number MFCD00135121
InChI Key YBVOPUOUKFHORP-UHFFFAOYSA-M
Synonym (Heptafluoropropyl)phenyliodonium Trifluoromethanesulfonate
PubChem CID 2760327
IUPAC Name 1,1,2,2,3,3,3-heptafluoropropyl(phenyl)iodanium;trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)[I+]C(C(C(F)(F)F)(F)F)(F)F.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 522.10
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Perfluoropropyl)phenyliodonium Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.