accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7778-85-0

IUPAC Name: 1,2-dimethoxypropane
Synonyms: 1,2-dimethoxypropane
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: LEEANUDEDHYDTG-UHFFFAOYNA-N
SMILES: COCC(C)OC
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results