accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOCH3H2C

CAS RN 7779-31-9

IUPAC Name: 3,3,5-trimethylcyclohexyl 2-methylprop-2-enoate
Synonyms: 3,3,5-trimethylcyclohexyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 210.32
Molecular Formula: C13H22O2
InChi Key: DABQKEQFLJIRHU-UHFFFAOYNA-N
SMILES: CC1CC(CC(C)(C)C1)OC(=O)C(C)=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results