accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOCH3

CAS RN 7779-75-1

IUPAC Name: 2-methylpropyl 3-oxobutanoate
Synonyms: 2-methylpropyl 3-oxobutanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: ZYXNLVMBIHVDRH-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)CC(C)=O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results