accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3CH3(E/Z)

CAS RN 7779-81-9

IUPAC Name: 2-methylpropyl 2-methylbut-2-enoate
Synonyms: isobutyl angelate
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: XDEGQMQKHFPBEW-UHFFFAOYSA-N
SMILES: CC=C(C)C(=O)OCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results