Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Angelate 98.0+%, TCI America™
SDP

Catalog No. A112725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A112725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A112725ML Supplier TCI America Supplier No. A112725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7779-81-9
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00063646
InChI Key XDEGQMQKHFPBEW-YVMONPNESA-N
Synonym Angelic Acid Isobutyl Ester, (Z)-2-Methyl-2-butenoic Acid Isobutyl Ester, 2-Methylisocrotonic Acid Isobutyl Ester, Isobutyl (Z)-2-Methyl-2-butenoate
PubChem CID 5367807
IUPAC Name 2-methylpropyl (Z)-2-methylbut-2-enoate
SMILES CC=C(C)C(=O)OCC(C)C

Specifications

Color Colorless
Boiling Point 177°C
Quantity 25 mL
UN Number 3272
Formula Weight 156.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Angelate
EINECSNumber (2)-3282
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.