accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7779-90-0

IUPAC Name: trizinc(2+) diphosphate
Synonyms: zinc phosphate
Molecular Weight (g/mol): 386.08
Molecular Formula: O8P2Zn3
InChi Key: LRXTYHSAJDENHV-UHFFFAOYSA-H
SMILES: [Zn++].[Zn++].[Zn++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)