accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C

CAS RN 778-22-3

IUPAC Name: (2-phenylpropan-2-yl)benzene
Synonyms: (2-phenylpropan-2-yl)benzene
Molecular Weight (g/mol): 196.29
Molecular Formula: C15H16
InChi Key: MILSYCKGLDDVLM-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results