Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diphenylpropane 97%, Thermo Scientific™

Catalog No. AC117400050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC117400050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC117400050 Supplier Thermo Scientific Chemicals Supplier No. 117400050

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 778-22-3
Molecular Formula C15H16
Molecular Weight (g/mol) 196.293
InChI Key MILSYCKGLDDVLM-UHFFFAOYSA-N
Synonym 2,2-diphenylpropane, unii-ov5r1azu4n, propane, 2,2-diphenyl, dimethyldiphenylmethane, ov5r1azu4n, propane-2,2-diyldibenzene, benzene, 1,1'-1-methylethylidene bis, 2-phenylpropan-2-yl benzene, 2',2'-diphenylpropane, 22dpp
PubChem CID 13065
IUPAC Name 2-phenylpropan-2-ylbenzene
SMILES CC(C)(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.99
Infrared Spectrum Authentic
Refractive Index 1.5690 to 1.5710 (20°C, 589nm)
Linear Formula CH3C(C6H5)2CH3
Quantity 5 g
Beilstein 05, 614
Formula Weight 196.29
Percent Purity ≥96.0%
Physical Form Liquid After Melting
Chemical Name or Material 2, 2-Diphenylpropane, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.