accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 7782-50-5

IUPAC Name: dichlorine
Synonyms: chlorine Cl2 molecular chlorine dichlorine
Molecular Weight (g/mol): 70.9
Molecular Formula: Cl2
InChi Key: KZBUYRJDOAKODT-UHFFFAOYSA-N
SMILES: ClCl
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (1)
Quantity 
  • (3)
  • (1)
  • (2)
Grade 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)