Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Chlorine Water
SCI_ED

Catalog No. S25247 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25247 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25247

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7782-50-5 , 7732-18-5
Molecular Formula Cl2
Molecular Weight (g/mol) 70.9
InChI Key KZBUYRJDOAKODT-UHFFFAOYSA-N
Synonym chlorine, bertholite, chloor, cloro, chlorine gas, chlorine mol., chlore french, cloro italian, chloor dutch, chlor german
PubChem CID 24526
ChEBI CHEBI:29310
IUPAC Name molecular chlorine
SMILES ClCl

Specifications

Packaging Solution
Quantity 1 L
Chemical Name or Material Chlorine Water
Safety handling Check Alert storage code Red
ShelfLife 6 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.