accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OMgOHPOOO

CAS RN 7782-75-4

IUPAC Name: magnesium(2+) trihydrate hydrogen phosphate
Synonyms: magnesium(2+) trihydrate hydrogen phosphate
Molecular Weight (g/mol): 174.33
Molecular Formula: H7MgO7P
InChi Key: OKIWLDVQGKRUNR-UHFFFAOYSA-L
SMILES: O.O.O.[Mg++].OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (1)
  • (2)
Grade 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)