Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Phosphate Dibasic Trihydrate, MP Biomedicals

Catalog No. ICN15530383 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15530383 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15530383 Supplier MP Biomedicals, Inc Supplier No. 0215530383

Chemical Identifiers

CAS 7782-75-4
Molecular Formula H7MgO7P
Molecular Weight (g/mol) 174.33
MDL Number MFCD00150390
InChI Key OKIWLDVQGKRUNR-UHFFFAOYSA-L
Synonym magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
PubChem CID 16204530
IUPAC Name magnesium(2+) trihydrate hydrogen phosphate
SMILES O.O.O.[Mg++].OP([O-])([O-])=O

Specifications

Quantity 250 g
Formula Weight 174.3
Chemical Name or Material Magnesium Phosphate Dibasic Trihydrate
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.