accessibility menu, dialog, popup

UserName

Viewing profile as user:
HgONOOHOHOHHONOOHg

CAS RN 7782-86-7

IUPAC Name: dimercury(1+) dihydrate dinitrate
Synonyms: dimercury(1+) dihydrate dinitrate
Molecular Weight (g/mol): 561.22
Molecular Formula: H4Hg2N2O8
InChi Key: LSABZDVKJBWCBE-UHFFFAOYSA-N
SMILES: O.O.[Hg+].[Hg+].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Grade 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)