CAS RN 7784-27-2
Aluminum Nitrate Nonahydrate (Cryst./Certified ACS), Fisher Chemical™
CAS: 7784-27-2 Molecular Formula: AlH18N3O18 Molecular Weight (g/mol): 375.129 MDL Number: MFCD00149132 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N Synonym: aluminum nitrate nonahydrate,aluminum trinitrate nonahydrate,unii-8mc6621v1h,aluminum iii nitrate, nonahydrate 1:3:9,nitric acid aluminum salt,nitric acid, aluminum salt, nonahydrate 8ci,9ci,aluminum nitrate, acs,aluminum 3+ nonahydrate trinitronate,aluminum nitrate nonahydrate, low mercury, puratronic PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Aluminum nitrate nonahydrate, ACS, 98.0-102.0%
CAS: 7784-27-2 Molecular Formula: AlH18N3O18 Molecular Weight (g/mol): 375.129 MDL Number: MFCD00003419 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N Synonym: aluminum nitrate nonahydrate,aluminum trinitrate nonahydrate,unii-8mc6621v1h,aluminum iii nitrate, nonahydrate 1:3:9,nitric acid aluminum salt,nitric acid, aluminum salt, nonahydrate 8ci,9ci,aluminum nitrate, acs,aluminum 3+ nonahydrate trinitronate,aluminum nitrate nonahydrate, low mercury, puratronic PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Aluminum nitrate nonahydrate, 99+%, for analysis
CAS: 7784-27-2 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N Synonym: aluminum nitrate nonahydrate,aluminum trinitrate nonahydrate,unii-8mc6621v1h,aluminum iii nitrate, nonahydrate 1:3:9,nitric acid aluminum salt,nitric acid, aluminum salt, nonahydrate 8ci,9ci,aluminum nitrate, acs,aluminum 3+ nonahydrate trinitronate,aluminum nitrate nonahydrate, low mercury, puratronic PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis)
CAS: 7784-27-2 Molecular Formula: AlH18N3O18 Molecular Weight (g/mol): 375.129 MDL Number: MFCD00003419 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N Synonym: aluminum nitrate nonahydrate,aluminum trinitrate nonahydrate,unii-8mc6621v1h,aluminum iii nitrate, nonahydrate 1:3:9,nitric acid aluminum salt,nitric acid, aluminum salt, nonahydrate 8ci,9ci,aluminum nitrate, acs,aluminum 3+ nonahydrate trinitronate,aluminum nitrate nonahydrate, low mercury, puratronic PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Aluminum nitrate nonahydrate, 98%
CAS: 7784-27-2 Molecular Formula: AlH18N3O18 Molecular Weight (g/mol): 375.129 MDL Number: MFCD00003419 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N Synonym: aluminum nitrate nonahydrate,aluminum trinitrate nonahydrate,unii-8mc6621v1h,aluminum iii nitrate, nonahydrate 1:3:9,nitric acid aluminum salt,nitric acid, aluminum salt, nonahydrate 8ci,9ci,aluminum nitrate, acs,aluminum 3+ nonahydrate trinitronate,aluminum nitrate nonahydrate, low mercury, puratronic PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
VeriSpec™ Aluminum (Al) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical
CAS: 7784-27-2 Molecular Formula: AlH18N3O18 Molecular Weight (g/mol): 375.129 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3
CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol
Aluminum nitrate nonahydrate, 99.999%, (trace metal basis)
CAS: 7784-27-2 Molecular Formula: AlN3O9·9H2O Molecular Weight (g/mol): 375.13 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N Synonym: aluminum nitrate nonahydrate,aluminum trinitrate nonahydrate,unii-8mc6621v1h,aluminum iii nitrate, nonahydrate 1:3:9,nitric acid aluminum salt,nitric acid, aluminum salt, nonahydrate 8ci,9ci,aluminum nitrate, acs,aluminum 3+ nonahydrate trinitronate,aluminum nitrate nonahydrate, low mercury, puratronic PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml
CAS: 7784-27-2 Molecular Formula: AlH18N3O18 Molecular Weight (g/mol): 375.129 MDL Number: MFCD00134029 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Aluminum nitrate nonahydrate, ACS reagent, Thermo Scientific Chemicals
CAS: 7784-27-2 Molecular Formula: AlN3O9·9H2O Molecular Weight (g/mol): 375.13 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N Synonym: aluminum nitrate nonahydrate,aluminum trinitrate nonahydrate,unii-8mc6621v1h,aluminum iii nitrate, nonahydrate 1:3:9,nitric acid aluminum salt,nitric acid, aluminum salt, nonahydrate 8ci,9ci,aluminum nitrate, acs,aluminum 3+ nonahydrate trinitronate,aluminum nitrate nonahydrate, low mercury, puratronic PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Aluminum Nitrate, Nonahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
CAS: 7784-27-2
Aluminum ICP Standard, 10,000 ppm Al in 5% HCl, Ricca Chemical
CAS: 7784-27-2 Molecular Formula: AlH18N3O18 Molecular Weight (g/mol): 375.129 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Aluminum AA Standard, 1000 ppm Al in 3% HNO3, Ricca Chemical
CAS: 7784-27-2 Molecular Formula: AlH18N3O18 Molecular Weight (g/mol): 375.129 InChI Key: SWCIQHXIXUMHKA-UHFFFAOYSA-N PubChem CID: 24567 IUPAC Name: aluminum;trinitrate;nonahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
- 1
- 2