Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Nitrate, Nonahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18608531 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-531 50 kg Fiber Drum
18-608-525 25 g Poly Pail
18-608-526 125 g Amber Glass Bottle
18-608-527 500 g Amber Glass Bottle
18-608-528 2.5 kg Amber Glass Bottle
18-608-529 12 kg Amber Glass Bottle
18-608-530 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-608-531 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A109550KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7784-27-2

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 50 kg
Molecular Weight (g/mol) 375.13
Percent Purity 98 to 102%
Grade Reagent
Packaging Fiber Drum
Chemical Name or Material Aluminum Nitrate, Nonahydrate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.