accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3NH3H2OH2OH2OH2OH2OH2ONiOHSOOOOHSOOO

CAS RN 7785-20-8

IUPAC Name: nickel(2+) diamine hexahydrate dihydrogen sulfate
Synonyms: nickel(2+) diamine hexahydrate dihydrogen sulfate
Molecular Weight (g/mol): 394.97
Molecular Formula: H20N2NiO14S2
InChi Key: ZZPAXHDZFDUTLY-UHFFFAOYSA-L
SMILES: N.N.O.O.O.O.O.O.[Ni++].OS([O-])(=O)=O.OS([O-])(=O)=O
Quantity 
  • (3)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)