Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Ammonium Sulfate Hexahydrate (Crystalline/Certified), Fisher Chemical

Catalog No. N48500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
N48-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N48-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7785-20-8
Molecular Formula H8N2NiO8S · 6 H2O
MDL Number MFCD00150260
Synonym Nickel (II) Ammonium Sulfate

Specifications

Color Blue-Green
pH 4.6
Physical Form Solid
Quantity 500 g
Linear Formula Ni(NH4)2(SO4)2·6H2O
Formula Weight 395
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Density 1.9g/cm³
Chemical Name or Material Nickel Ammonium Sulfate Hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.