accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrAg

CAS RN 7785-23-1

IUPAC Name: silver(1+) bromide
Synonyms: silver bromide
Molecular Weight (g/mol): 187.77
Molecular Formula: AgBr
InChi Key: ADZWSOLPGZMUMY-UHFFFAOYSA-M
SMILES: [Br-].[Ag+]
Molecular Weight (g/mol) 
  • (6)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)