Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Bromide, Granular, Purified, 99%, Spectrum™ Chemical
SDP

Catalog No. 18613120 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
125 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-120 125 g Amber Glass Bottle
18-613-118 25 g Amber Glass Bottle
18-613-119 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-613-120 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1048125GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7785-23-1

Specifications

Melting Point 432°C
Color Yellow
CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 125 g
Molecular Formula AgBr
Linear Formula AgBr
Synonym Bromargyrite
IUPAC Name Silver(1+) bromide
Molecular Weight (g/mol) 187.8
Formula Weight 187.77 g/mol
Percent Purity 99%
Grade Purified
Odor Odorless
Packaging Amber Glass Bottle
Appearance Yellowish Powder
Chemical Name or Material Silver Bromide, Granular
Show More Show Less
Recommended Storage Light Sensitive, Store in a tightly closed container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.