accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH2HO

CAS RN 7786-61-0

IUPAC Name: 4-ethenyl-2-methoxyphenol
Synonyms: 2-methoxy-4-vinyl-phenol vinyl guaiacol
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: YOMSJEATGXXYPX-UHFFFAOYSA-N
SMILES: COC1=CC(C=C)=CC=C1O
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)