Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-4-vinylphenol, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015524 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5524 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5524 Supplier MilliporeSigma Supelco Supplier No. 89189100MG
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 7786-61-0
Density 1.11 g/mL (at 25°C)
Flash Point 113°C - closed cup
Physical Form Neat
Linear Formula CH3 OC6H3(CH=CH2)OH
MDL Number MFCD00015437
Boiling Point 224°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 150.17
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.