accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77866-58-1

CH3OH

CAS RN 77866-58-1

IUPAC Name: 4-propylcyclohexan-1-ol
Synonyms: 4-propylcyclohexan-1-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: YVPZFPKENDZQEJ-UHFFFAOYSA-N
SMILES: CCCC1CCC(O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results