Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Propylcyclohexanol 95.0+%, TCI America™
SDP

Catalog No. P18821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18821G Supplier TCI America Supplier No. P18821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77866-58-1
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00091618,MFCD11114411
InChI Key YVPZFPKENDZQEJ-UHFFFAOYSA-N
PubChem CID 2775094
IUPAC Name 4-propylcyclohexan-1-ol
SMILES CCCC1CCC(O)CC1

Specifications

Color Yellow
Boiling Point 208°C
Quantity 1 g
Formula Weight 142.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-4-Propylcyclohexanol
EINECSNumber (3)-2310
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.