accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7787-49-7

IUPAC Name: beryllium(2+) difluoride
Synonyms: beryllium fluoride
Molecular Weight (g/mol): 47.01
Molecular Formula: BeF2
InChi Key: JZKFIPKXQBZXMW-UHFFFAOYSA-L
SMILES: [Be++].[F-].[F-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results