Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beryllium Fluoride 98.5+%, Thermo Scientific™

Catalog No. AC197010250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC197010250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC197010250 Supplier Thermo Scientific Chemicals Supplier No. 197010250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-49-7
Molecular Formula BeF2
Molecular Weight (g/mol) 47.009
InChI Key JZKFIPKXQBZXMW-UHFFFAOYSA-L
Synonym beryllium fluoride, beryllium difluoride, beryllium fluoride be2f4, hsdb 355, beryllium fluoride beryllium and beryllium compounds, fluoroberyllate, berylliumfluorid, be2f4
PubChem CID 24589
ChEBI CHEBI:49499
IUPAC Name beryllium;difluoride
SMILES [Be+2].[F-].[F-]

Specifications

Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Powder
Quantity 25 g
Linear Formula BeF2
Merck Index 15, 1169
Formula Weight 47
Percent Purity ≥98.50%
Density 1.98
Chemical Name or Material Beryllium fluoride, 98.5+%
Show More Show Less
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.