accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OKCrOSOOOOSOOO

CAS RN 7788-99-0

IUPAC Name: chromium(3+) potassium dodecahydrate disulfate
Synonyms: chrome alum
Molecular Weight (g/mol): 499.39
Molecular Formula: CrH24KO20S2
InChi Key: ZFVHBEKVAITXHW-UHFFFAOYSA-J
SMILES: O.O.O.O.O.O.O.O.O.O.O.O.[K+].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (16)
  • (4)
  • (7)
Quantity 
  • (7)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (9)
Physical Form 
  • (2)
  • (8)
  • (7)
  • (5)
Grade 
  • (3)
  • (5)
  • (3)
  • (1)
  • (4)
Percent Purity 
  • (4)
  • (3)
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (1)
  • (2)
  • (2)
  • (4)
  • (7)
  • (11)
  • (1)
  • (2)
  • (16)
  • (4)
  • (7)
  • (7)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (9)
  • (2)
  • (8)
  • (7)
  • (5)
  • (3)
  • (5)
  • (3)
  • (1)
  • (4)
  • (4)
  • (3)
  • (3)
  • (5)