Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Potassium Sulfate, Dodecahydrate, MP Biomedicals™

Catalog No. MP219403105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219403105 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219403105 Supplier MP Biomedicals, Inc Supplier No. 0219403105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chromium potassium sulfate is the potassium double sulfate of chromium.

Chromium alum is used in the tanning of leather. It was also used in gelatine emulsions in photographic film as a hardener.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7788-99-0
Molecular Formula CrH24KO20S2
Molecular Weight (g/mol) 499.386
InChI Key ZFVHBEKVAITXHW-UHFFFAOYSA-J
Synonym chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
PubChem CID 24596
IUPAC Name potassium;chromium(3+);disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]

Specifications

Melting Point 89°C (lit.)
Physical Form Crystals
Quantity 5 g
Assay Percent Range 98.0-102.0%
Density 1.89 g/cm3 (at 20°C)
Solubility Information Soluble in water.
Formula Weight 499.4g/mol
Grade ACS
Recommended Storage Store at Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.