accessibility menu, dialog, popup

UserName

Viewing profile as user:
ICs

CAS RN 7789-17-5

IUPAC Name: caesium(1+) iodide
Synonyms: cesium iodide|caesium iodide
Molecular Weight (g/mol): 259.81
Molecular Formula: CsI
InChi Key: XQPRBTXUXXVTKB-UHFFFAOYSA-M
SMILES: [I-].[Cs+]
Quantity 
  • (3)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (5)
  • (3)
Physical Form 
  • (4)
  • (2)
  • (6)
  • (2)
Grade 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (13)
  • (3)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (5)
  • (3)
  • (4)
  • (2)
  • (6)
  • (2)
  • (2)
  • (3)
  • (2)