Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Iodide, 99.9%, MP Biomedicals

Catalog No. ICN218738 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218738 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218738 Supplier MP Biomedicals, Inc Supplier No. 0521873810

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7789-17-5
Molecular Formula CsI
Molecular Weight (g/mol) 259.81
MDL Number MFCD00010962
InChI Key XQPRBTXUXXVTKB-UHFFFAOYSA-M
Synonym cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
PubChem CID 24601
IUPAC Name caesium(1+) iodide
SMILES [I-].[Cs+]

Specifications

Quantity 10 g
Formula Weight 259.81
Chemical Name or Material Cesium Iodide, 99.9%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.