CAS RN 7789-24-4
Lithium fluoride, 98.5%
CAS: 7789-24-4 Molecular Formula: FLi Molecular Weight (g/mol): 25.94 MDL Number: MFCD00011090 InChI Key: PQXKHYXIUOZZFA-UHFFFAOYSA-M Synonym: lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif PubChem CID: 224478 SMILES: [Li+].[F-]
Lithium fluoride, 99.98%, extra pure
CAS: 7789-24-4 Molecular Formula: FLi Molecular Weight (g/mol): 25.94 MDL Number: MFCD00011090 InChI Key: PQXKHYXIUOZZFA-UHFFFAOYSA-M Synonym: lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif PubChem CID: 224478 SMILES: [Li+].[F-]
Lithium fluoride, 97%
CAS: 7789-24-4 Molecular Formula: FLi Molecular Weight (g/mol): 25.94 MDL Number: MFCD00011090 InChI Key: PQXKHYXIUOZZFA-UHFFFAOYSA-M Synonym: lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif PubChem CID: 224478 IUPAC Name: lithium;fluoride SMILES: [Li+].[F-]
Lithium fluoride, 99.85% (metals basis)
CAS: 7789-24-4 Molecular Formula: FLi Molecular Weight (g/mol): 25.94 MDL Number: MFCD00011090 InChI Key: PQXKHYXIUOZZFA-UHFFFAOYSA-M Synonym: lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif PubChem CID: 224478 IUPAC Name: lithium;fluoride SMILES: [Li+].[F-]
Lithium fluoride, ultra dry, 99.99% (metals basis)
CAS: 7789-24-4 Molecular Formula: FLi Molecular Weight (g/mol): 25.94 MDL Number: MFCD00011090 InChI Key: PQXKHYXIUOZZFA-UHFFFAOYSA-M Synonym: lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif PubChem CID: 224478 SMILES: [Li+].[F-]
Lithium fluoride, 97%, for analysis
CAS: 7789-24-4 Molecular Formula: FLi Molecular Weight (g/mol): 25.94 MDL Number: MFCD00011090 InChI Key: PQXKHYXIUOZZFA-UHFFFAOYSA-M Synonym: lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif PubChem CID: 224478 SMILES: [Li+].[F-]
Lithium fluoride, Puratronic™, 99.99% (metals basis)
CAS: 7789-24-4 Molecular Formula: FLi Molecular Weight (g/mol): 25.94 MDL Number: MFCD00011090 InChI Key: PQXKHYXIUOZZFA-UHFFFAOYSA-M Synonym: lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif PubChem CID: 224478 SMILES: [Li+].[F-]
Lithium fluoride, 99.98% (metals basis)
CAS: 7789-24-4 Molecular Formula: FLi Molecular Weight (g/mol): 25.94 MDL Number: MFCD00011090 InChI Key: PQXKHYXIUOZZFA-UHFFFAOYSA-M Synonym: lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif PubChem CID: 224478 SMILES: [Li+].[F-]
Lithium fluoride, Optical Grade, 99.5+%, Thermo Scientific Chemicals
CAS: 7789-24-4 Molecular Formula: FLi Molecular Weight (g/mol): 25.94 MDL Number: MFCD00011090 InChI Key: PQXKHYXIUOZZFA-UHFFFAOYSA-M Synonym: lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif PubChem CID: 224478 SMILES: [Li+].[F-]
Sigma Aldrich Lithium fluoride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lithium fluoride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lithium fluoride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lithium fluoride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lithium fluoride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Lithium Fluoride, Optical Grade, 99.8%, MP Biomedicals
CAS: 7789-24-4 Molecular Formula: FLi Molecular Weight (g/mol): 25.94 MDL Number: MFCD00011090 InChI Key: PQXKHYXIUOZZFA-UHFFFAOYSA-M Synonym: lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif PubChem CID: 224478 IUPAC Name: lithium(1+) fluoride SMILES: [Li+].[F-]