Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium fluoride

Catalog No. 502891476
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-1476 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-289-1476 Supplier Sigma Aldrich Supplier No. 20364525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium fluoride
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1673°C
Melting Point 845°C
CAS 7789-24-4
Quantity 25 g
Molecular Formula FLi
Linear Formula LiF
MDL Number MFCD00011090
Synonym Fluorolithium
Molecular Weight (g/mol) 25.94
Percent Purity ≥99.98% trace metals basis
Density 2.64 g/mL (at 25°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.