accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7789-46-0

IUPAC Name: λ²-iron(2+) dibromide
Synonyms: iron(II) bromide
Molecular Weight (g/mol): 215.65
Molecular Formula: Br2Fe
InChi Key: GYCHYNMREWYSKH-UHFFFAOYSA-L
SMILES: [Fe++].[Br-].[Br-]
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)