Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog No. AA1454103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1454103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1454103 Supplier Thermo Scientific Chemicals Supplier No. 01454103
Add to Cart

missing translation for 'validMainframeMsg'

Used as a polymerization catalyst. It is used in water treatment, chemical analysis and in ultra high purity for certain crystal growth applications.

Iron(II) bromide is used as a polymerization catalyst. It is used in water treatment, chemical analysis and in ultra high purity for certain crystal growth applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-46-0
Molecular Formula Br2Fe
Molecular Weight (g/mol) 215.65
MDL Number MFCD00016081
InChI Key GYCHYNMREWYSKH-UHFFFAOYSA-L
Synonym iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
PubChem CID 82240
IUPAC Name iron(2+);dibromide
SMILES [Fe++].[Br-].[Br-]

Specifications

Melting Point 684°C
Physical Form Beads
Quantity 1 g
Packaging Ampouled under argon
Sensitivity Air sensitive; Hygroscopic
Specific Gravity 4.636
Density 4.636
UN Number UN3260
Solubility Information Soluble in water,THF,methanol,ethanol.
Formula Weight 215.67
Percent Purity 99.995%
Chemical Name or Material Iron(II) bromide
Show More Show Less
Hazard Category H314-H318
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 232-168-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.