accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7789-68-6

IUPAC Name: titanium(4+) tetrabromide
Synonyms: titanium tetrabromide
Molecular Weight (g/mol): 367.48
Molecular Formula: Br4Ti
InChi Key: UBZYKBZMAMTNKW-UHFFFAOYSA-J
SMILES: [Ti+4].[Br-].[Br-].[Br-].[Br-]
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)