accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7789-77-7

IUPAC Name: calcium dihydrate hydrogen phosphate
Synonyms: dibasic calcium phosphate phosphoric acid, calcium salt (1:1), dihydrate CaHPO4.2H2O dicalcium phosphate dibasic calcium phosphate dihydrate
Molecular Weight (g/mol): 172.09
Molecular Formula: CaH5O6P
InChi Key: XAAHAAMILDNBPS-UHFFFAOYSA-L
SMILES: O.O.[Ca++].OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
  • (1)
  • (4)
Physical Form 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (4)
  • (2)
  • (3)
  • (6)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
  • (1)