Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate Dibasic Dihydrate (Powder/USP/FCC), Fisher Chemical

Catalog No. C129212 Shop All Fisherbrand Products
Change view
Click to view available options
Poly Bottle; 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
C129-212 Poly Bottle; 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. C129-212

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color White
CAS 7789-77-7
pH 7.5
Physical Form Solid
Quantity 2.5 kg
Molecular Formula CaH5O6P
MDL Number MFCD00149621
Synonym calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen
InChI Key XAAHAAMILDNBPS-UHFFFAOYSA-L
SMILES O.O.OP(=O)([O-])[O-].[Ca+2]
IUPAC Name calcium;hydrogen phosphate;dihydrate
Molecular Weight (g/mol) 172.086
PubChem CID 104805
ChEBI CHEBI:4496
Formula Weight 172.09
Percent Purity 98.0 to 105.0% (USP)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (FCC/USP), Pass Test (USP)
Loss on Ignition 24.5 to 26.5% (FCC), 24.5 to 26.5% (FCC/USP), 24.5 to 26.5% (USP)
Packaging Poly Bottle
Chemical Name or Material Calcium Phosphate Dibasic Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.