accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaOMoOOO

CAS RN 7789-82-4

IUPAC Name: calcium dioxomolybdenumbis(olate)
Synonyms: calcium molybdate
Molecular Weight (g/mol): 200.02
Molecular Formula: CaMoO4
InChi Key: BIOOACNPATUQFW-UHFFFAOYSA-N
SMILES: [Ca++].[O-][Mo]([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (3)
  • (1)
  • (2)