Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium molybdenum oxide, 99.8%, Thermo Scientific™

Catalog No. AA4186522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4186522 100 g
AA4186514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4186522 Supplier Thermo Scientific Chemicals Supplier No. 4186522

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-82-4
Molecular Formula CaMoO4
Molecular Weight (g/mol) 200.02
MDL Number MFCD00015976
InChI Key BIOOACNPATUQFW-UHFFFAOYSA-N
Synonym calcium molybdate, molybdate, calcium, calcium molybdate vi, unii-q03c83v2an, calcium molybdenum oxide, molybdic acid h2moo4 , calcium salt 1:1, molybdate moo42-, calcium 1:1 , t-4, molybdate moo42-, calcium 1:1 , beta-4, calcium dioxido dioxo molybdenum, calcium 2+ molybdate
PubChem CID 24620
IUPAC Name calcium;dioxido(dioxo)molybdenum
SMILES [Ca++].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 965°C
Color White
Physical Form Powder
Quantity 100 g
Merck Index 14,1681
Solubility Information Soluble in mineral acids. Insoluble in water,ethanol,ethyl ether.
Formula Weight 200.01
Percent Purity 99.8%
Odor Odorless
Density 4.45 g/mL
Chemical Name or Material Calcium molybdenum oxide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-192-9
RTECSNumber EW2975000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.