accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3OCH3O

CAS RN 779-90-8

IUPAC Name: 1-(3,5-diacetylphenyl)ethan-1-one
Synonyms: 1-(3,5-diacetylphenyl)ethanone
Molecular Weight (g/mol): 204.23
Molecular Formula: C12H12O3
InChi Key: HSOAIPRTHLEQFI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=CC(=C1)C(C)=O)C(C)=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)