Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Triacetylbenzene 98%, Thermo Scientific™

Catalog No. AC139231000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC139231000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC139231000 Supplier Thermo Scientific Chemicals Supplier No. 139231000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 779-90-8
Molecular Formula C12H12O3
Molecular Weight (g/mol) 204.225
InChI Key HSOAIPRTHLEQFI-UHFFFAOYSA-N
Synonym 1,3,5-triacetylbenzene, 1,1',1-benzene-1,3,5-triyl triethanone, 1,3,5,triacetylbenzene, benzene, 1,3,5-triacetyl, 1-3,5-diacetylphenyl ethanone, 1,5-triacetylbenzene, benzene,3,5-triacetyl, acmc-1bc4n, cambridge id 6741393, ethanone, 1,1',1-1,3,5-benzenetriyl tris
PubChem CID 69904
IUPAC Name 1-(3,5-diacetylphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1)C(=O)C)C(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 160°C to 164°C
Color Beige to Cream
Infrared Spectrum Authentic
Linear Formula C6H3(COCH3)3
Quantity 100 g
Beilstein 07, 866
Formula Weight 204.22
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3, 5-Triacetylbenzene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.