accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7790-21-8

IUPAC Name: potassium periodate
Synonyms: potassium periodate
Molecular Weight (g/mol): 230
Molecular Formula: IKO4
InChi Key: FJVZDOGVDJCCCR-UHFFFAOYSA-M
SMILES: [K+].[O-][I](=O)(=O)=O
Quantity 
  • (7)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
Molecular Weight (g/mol) 
  • (13)
  • (5)
Grade 
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (6)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (10)
  • (7)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (13)
  • (5)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (6)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)